accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3

CAS RN 29665-59-6

IUPAC Name: 4-dodecyl-2-methylphenol
Synonyms: 4-dodecyl-2-methylphenol
Molecular Weight (g/mol): 276.46
Molecular Formula: C19H32O
InChi Key: RJLWSJUUDKAMPQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC(C)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results