accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2ClHN

CAS RN 29671-92-9

IUPAC Name: hydrogen aminomethanecarbonimidoyl chloride chloride
Synonyms: hydrogen aminomethanecarbonimidoyl chloride chloride
Molecular Weight (g/mol): 114.96
Molecular Formula: CH4Cl2N2
InChi Key: FUQFHOLPJJETAP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(Cl)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results