accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONBr

CAS RN 29681-44-5

IUPAC Name: methyl 5-bromopyridine-3-carboxylate
Synonyms: methyl 5-bromopyridine-3-carboxylate
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: AAJZXPWBILCHAW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CC(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)