accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrN

CAS RN 29682-15-3

IUPAC Name: methyl 5-bromopyridine-2-carboxylate
Synonyms: methyl 5-bromopyridine-2-carboxylate
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: JEURNBCYNWNADN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)C=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)