accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNS(E)

CAS RN 29683-84-9

IUPAC Name: (E)-N-[(thiophen-2-yl)methylidene]hydroxylamine
Synonyms: (E)-N-(thiophen-2-ylmethylidene)hydroxylamine
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: GASLBPLHYRZLLT-GQCTYLIASA-N
SMILES: O\N=C\C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results