accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2969-81-5

IUPAC Name: ethyl 4-bromobutanoate
Synonyms: ethyl 4-bromobutanoate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: XBPOBCXHALHJFP-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCBr
Boiling Point 
  • (3)
  • (1)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (9)