accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3ONN

CAS RN 297-97-2

IUPAC Name: O,O-diethyl O-pyrazin-2-yl phosphorothioate
Synonyms: thionazin
Molecular Weight (g/mol): 248.24
Molecular Formula: C8H13N2O3PS
InChi Key: IRVDMKJLOCGUBJ-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=CN=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results