accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29700-22-9

IUPAC Name: 4-[(1E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: oxyresveratrol hydroxyresveratrol
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: PDHAOJSHSJQANO-OWOJBTEDSA-N
SMILES: OC1=CC=C(\C=C\C2=CC(O)=CC(O)=C2)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results