accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFOHOHOHHO

CAS RN 29702-43-0

IUPAC Name: 2-fluoro-3,4,5,6-tetrahydroxyhexanal
Synonyms: 2-fluoro-3,4,5,6-tetrahydroxyhexanal
Molecular Weight (g/mol): 182.15
Molecular Formula: C6H11FO5
InChi Key: AOYNUTHNTBLRMT-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C(F)C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results