accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnSCH3S

CAS RN 29726-21-4

IUPAC Name: 5-methylbenzo[d]1,3-dithia-2-zincacyclopentane-2,2-bis(ylium)-1,3-diuide
Synonyms: 5-methylbenzo[d]1,3-dithia-2-zincacyclopentane-2,2-bis(ylium)-1,3-diuide
Molecular Weight (g/mol): 219.63
Molecular Formula: C7H6S2Zn
InChi Key: WSPDPEGUBOEWME-UHFFFAOYSA-L
SMILES: [Zn++].CC1=CC=C([S-])C([S-])=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)