accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2973-75-3

IUPAC Name: 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
Synonyms: 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol): 309.94
Molecular Formula: C8H6Br2O3
InChi Key: WKLKGSHBXNPUDU-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=C(Br)C(Br)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results