accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrHO

CAS RN 2973-76-4

IUPAC Name: 3-bromo-4-hydroxy-5-methoxybenzaldehyde
Synonyms: 3-bromo-4-hydroxy-5-methoxybenzaldehyde
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: KLSHZDPXXKAHIJ-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(Br)=C1O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)