accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2973-78-6

IUPAC Name: 3-bromo-4-hydroxybenzaldehyde
Synonyms: 3-bromo-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: UOTMHAOCAJROQF-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C=C1Br
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)