accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOBr

CAS RN 2973-80-0

IUPAC Name: 2-bromo-5-hydroxybenzaldehyde
Synonyms: 2-bromo-5-hydroxybenzaldehyde
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: SCRQAWQJSSKCFN-UHFFFAOYSA-N
SMILES: OC1=CC(C=O)=C(Br)C=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)