accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29743-08-6

IUPAC Name: N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine
Synonyms: N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine
Molecular Weight (g/mol): 281.4
Molecular Formula: C19H23NO
InChi Key: DBOAVDSSZWDGTH-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)N=CC1=CC=C(OCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results