accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29753-26-2

IUPAC Name: 1-(4-aminophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(4-aminophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: XOPCHXSYQHXLHJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1C(=O)C=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results