accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOH3CSSH3C

CAS RN 298-04-4

IUPAC Name: O,O-diethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonothioate
Synonyms: frumin caswell no. 341 glebofos disyston rcra waste number P039 ethyl thiometon ethylthiometon B di-syston G demeton (substance) insyst-D
Molecular Weight (g/mol): 274.39
Molecular Formula: C8H19O2PS3
InChi Key: DOFZAZXDOSGAJZ-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCCSCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results