accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29823-18-5

IUPAC Name: ethyl 7-bromoheptanoate
Synonyms: ethyl 7-bromoheptanoate
Molecular Weight (g/mol): 237.14
Molecular Formula: C9H17BrO2
InChi Key: OOBFNDGMAGSNKA-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCBr
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)