accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29823-87-8

IUPAC Name: 8-(dimethylamino)octan-1-ol
Synonyms: 8-(dimethylamino)octan-1-ol
Molecular Weight (g/mol): 173.3
Molecular Formula: C10H23NO
InChi Key: CAKJEDBOIMYCHP-UHFFFAOYSA-N
SMILES: CN(C)CCCCCCCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results