accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 2984-50-1

IUPAC Name: 2-hexyloxirane
Synonyms: 2-hexyloxirane
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: NJWSNNWLBMSXQR-UHFFFAOYNA-N
SMILES: CCCCCCC1CO1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)