accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29840-57-1

IUPAC Name: ethyl 5-aminopentanoate hydrochloride
Synonyms: ethyl 5-aminopentanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: JELXJZASXLHKDV-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)CCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)