accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29843-90-1

IUPAC Name: neodymium(3+) tribromide
Synonyms: neodymium(III) bromide
Molecular Weight (g/mol): 383.95
Molecular Formula: Br3Nd
InChi Key: LBWLQVSRPJHLEY-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Nd+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results