accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOOH3C

CAS RN 29865-90-5

IUPAC Name: 3-hydroxy-4,5-dimethoxybenzaldehyde
Synonyms: 3-hydroxy-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: NVLTWXMZECWWPC-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(O)=C1OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)