accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29874-01-9

IUPAC Name: chloro[(3-chlorophenyl)methyl]magnesium
Synonyms: chloro[(3-chlorophenyl)methyl]magnesium
Molecular Weight (g/mol): 185.33
Molecular Formula: C7H6Cl2Mg
InChi Key: FXQFOXTUJOAWNN-UHFFFAOYSA-M
SMILES: Cl[Mg]CC1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results