accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29875-07-8

IUPAC Name: chloro[(4-methylphenyl)methyl]magnesium
Synonyms: chloro[(4-methylphenyl)methyl]magnesium
Molecular Weight (g/mol): 164.92
Molecular Formula: C8H9ClMg
InChi Key: SAKYCLWKOWGTFT-UHFFFAOYSA-M
SMILES: CC1=CC=C(C[Mg]Cl)C=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)