accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)(R)(R)(S)(S)(R)OO(S)(S)(R)(R)ONOOHOHHOHOHOHOHO

CAS RN 29883-15-6

IUPAC Name: 2-phenyl-2-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxan-2-yl]oxy}acetonitrile
Synonyms: amygdalin
Molecular Weight (g/mol): 457.43
Molecular Formula: C20H27NO11
InChi Key: XUCIJNAGGSZNQT-ARGBPYSKNA-N
SMILES: OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)