accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2989-98-2

IUPAC Name: (3-bromophenyl)urea
Synonyms: M-bromophenylurea
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: DHMRSMNEKFDABI-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results