accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3ClAu

CAS RN 29892-37-3

IUPAC Name: (methylsulfanyl)methane; chlorogold
Synonyms: chloro(dimethyl sulfide)gold(I)
Molecular Weight (g/mol): 294.55
Molecular Formula: C2H6AuClS
InChi Key: YQALRAGCVWJXGB-UHFFFAOYSA-M
SMILES: Cl[Au].CSC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results