CAS RN 299-28-5
Calcium D-gluconate gel, 2.5% w/w aq. soln.
CAS: 299-28-5 Molecular Formula: C12H22CaO14 Molecular Weight (g/mol): 430.372 MDL Number: MFCD00064209 InChI Key: NEEHYRZPVYRGPP-IYEMJOQQSA-L Synonym: calcium gluconate,calcium d-gluconate,calciofon,calglucon,glucobiogen,ebucin,calcicol,calcipur,calglucol,dragocal PubChem CID: 9290 IUPAC Name: calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
Calcium Gluconate Monohydrate 98.0+%, TCI America™
CAS: 299-28-5 Molecular Formula: C12H22CaO14 Molecular Weight (g/mol): 430.372 MDL Number: MFCD00064209 InChI Key: NEEHYRZPVYRGPP-IYEMJOQQSA-L Synonym: calcium gluconate,calcium d-gluconate,calciofon,calglucon,glucobiogen,ebucin,calcicol,calcipur,calglucol,dragocal PubChem CID: 9290 IUPAC Name: calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
D-Gluconic acid, calcium salt, 99%
CAS: 299-28-5 Molecular Formula: C12H22CaO14 Molecular Weight (g/mol): 430.372 MDL Number: MFCD00064209 InChI Key: NEEHYRZPVYRGPP-IYEMJOQQSA-L Synonym: calcium gluconate,calcium d-gluconate,calciofon,calglucon,glucobiogen,ebucin,calcicol,calcipur,calglucol,dragocal PubChem CID: 9290 IUPAC Name: calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
Calcium Gluconate, Anhydrous, Powder, USP, 98-102%, Spectrum™ Chemical
CAS: 299-28-5 Molecular Formula: C12H22CaO14 Molecular Weight (g/mol): 430.37 InChI Key: NEEHYRZPVYRGPP-IYEMJOQQSA-L IUPAC Name: calcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) SMILES: [Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O