accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HNOHOHOHH3CCH3HNOHOHOHOHSOHOO

CAS RN 299-95-6

IUPAC Name: bis(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol); sulfuric acid
Synonyms: bis(isoproterenol); sulfuric acid
Molecular Weight (g/mol): 520.59
Molecular Formula: C22H36N2O10S
InChi Key: ZOLBALGTFCCTJF-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.CC(C)NCC(O)C1=CC=C(O)C(O)=C1.CC(C)NCC(O)C1=CC=C(O)C(O)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results