accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2991-28-8

IUPAC Name: 2,5-difluorobenzoic acid
Synonyms: 2,5-difluorobenzoic acid
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: LBQMIAVIGLLBGW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results