accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2991-84-6

IUPAC Name: nonafluorobutane-1-sulfonyl chloride
Synonyms: nonafluorobutane-1-sulfonyl chloride
Molecular Weight (g/mol): 318.54
Molecular Formula: C4ClF9O2S
InChi Key: IRFCLLARAUQTNK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)