accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOBr

CAS RN 29921-57-1

IUPAC Name: propan-2-yl 2-bromoacetate
Synonyms: isopropyl 2-bromoacetate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: JCWLEWKPXYZHGQ-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CBr
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)