accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiFFFFFFAs

CAS RN 29935-35-1

IUPAC Name: λ⁵-arsenic(5+) lithium(1+) hexafluoride
Synonyms: λ⁵-arsenic(5+) lithium(1+) hexafluoride
Molecular Weight (g/mol): 195.85
Molecular Formula: AsF6Li
InChi Key: YYPUQABLGAAHDV-UHFFFAOYSA-H
SMILES: [Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)