CAS RN 2994-71-0
IUPAC Name:
1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Synonyms:
1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Molecular Weight (g/mol):
300.05
Molecular Formula:
C6F12
InChi Key:
RBTROQHBNLSUTL-UHFFFAOYNA-N
SMILES:
FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals
Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals
CAS: 2994-71-0 Molecular Formula: C6F12 Molecular Weight (g/mol): 300.05 MDL Number: MFCD00013737 InChI Key: RBTROQHBNLSUTL-UHFFFAOYNA-N Synonym: Dodecafluorodimethylcyclobutane PubChem CID: 12789874 SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F