accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 299465-28-4

IUPAC Name: zinc(2+) dibromide
Synonyms: zinc bromide
Molecular Weight (g/mol): 225.19
Molecular Formula: Br2Zn
InChi Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES: [Zn++].[Br-].[Br-]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results