accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29949-85-7

CAS RN 29949-85-7

IUPAC Name: tris(3-chlorophenyl)phosphane
Synonyms: tris(3-chlorophenyl)phosphane
Molecular Weight (g/mol): 365.62
Molecular Formula: C18H12Cl3P
InChi Key: QAPGHLJQIVDTPT-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC=C1)P(C1=CC(Cl)=CC=C1)C1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results