accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3SiCH3H3CCH3Br

CAS RN 29955-12-2

IUPAC Name: [bromo(trimethylsilyl)methyl]trimethylsilane
Synonyms: [bromo(trimethylsilyl)methyl]trimethylsilane
Molecular Weight (g/mol): 239.3
Molecular Formula: C7H19BrSi2
InChi Key: MYXGYPFZVNSVCB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(Br)[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results