accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29956-99-8

IUPAC Name: 2,2,4-trimethyl-4-[(2,4,4-trimethylpentan-2-yl)disulfanyl]pentane
Synonyms: 2,2,4-trimethyl-4-[(2,4,4-trimethylpentan-2-yl)disulfanyl]pentane
Molecular Weight (g/mol): 290.57
Molecular Formula: C16H34S2
InChi Key: ZIMCZOLRXKPXLN-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)SSC(C)(C)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results