accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSH3CCH3BrBrBr

CAS RN 29957-59-3

IUPAC Name: (methylsulfanyl)methane; tribromoborane
Synonyms: boron tribromide; dimethyl sulfide
Molecular Weight (g/mol): 312.65
Molecular Formula: C2H6BBr3S
InChi Key: YNLFEVAOQLXINF-UHFFFAOYSA-N
SMILES: CSC.BrB(Br)Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results