accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2997-92-4

IUPAC Name: hydrogen 2-[2-(1-carbamimidoyl-1-methylethyl)diazen-1-yl]-2-methylpropanimidamide chloride
Synonyms: hydrogen 2-[2-(1-carbamimidoyl-1-methylethyl)diazen-1-yl]-2-methylpropanimidamide chloride
Molecular Weight (g/mol): 234.73
Molecular Formula: C8H19ClN6
InChi Key: QMYCJCOPYOPWTI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(N=NC(C)(C)C(N)=N)C(N)=N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)