accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONO

CAS RN 29976-53-2

IUPAC Name: ethyl 4-oxopiperidine-1-carboxylate
Synonyms: ethyl 4-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 171.2
Molecular Formula: C8H13NO3
InChi Key: LUBGFMZTGFXIIN-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCC(=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results