accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHC

CAS RN 29978-83-4

IUPAC Name: 2-(prop-2-yn-1-yloxy)benzaldehyde
Synonyms: 2-(prop-2-yn-1-yloxy)benzaldehyde
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: PEERMPFPCWSJLC-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OCC#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)