CAS RN 300-76-5
IUPAC Name:
1,2-dibromo-2,2-dichloroethyl dimethyl phosphate
Synonyms:
naled bromchlophos caswell no. 586 epa pesticide chemical code 034401 fosbrom ortho-dibrom bromex (insecticide) BRP hibrom dibromfos
Molecular Weight (g/mol):
380.78
Molecular Formula:
C4H7Br2Cl2O4P
InChi Key:
BUYMVQAILCEWRR-UHFFFAOYNA-N
SMILES:
COP(=O)(OC)OC(Br)C(Cl)(Cl)Br
1
–
1
of
1
results
1
Naled (Dibrom), SPEX CertiPrep™
CAS: 300-76-5 Molecular Formula: C4H7Br2Cl2O4P Molecular Weight (g/mol): 380.778 InChI Key: BUYMVQAILCEWRR-UHFFFAOYSA-N PubChem CID: 4420 ChEBI: CHEBI:38729 IUPAC Name: (1,2-dibromo-2,2-dichloroethyl) dimethyl phosphate SMILES: COP(=O)(OC)OC(C(Cl)(Cl)Br)Br