accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 300-85-6

IUPAC Name: 3-hydroxybutanoic acid
Synonyms: 3 hydroxybutyrate acid, 3-hydroxybutyric 3 hydroxybutyric acid 3-OH-butyric acid poly-β-hydroxybutyric acid β-hydroxybuttersaeure 3-hydroxybutyrate (substance) hydrxoy-butanoic acid 3 hba BHBA
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: WHBMMWSBFZVSSR-UHFFFAOYNA-N
SMILES: CC(O)CC(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (4)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (8)
  • (7)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (5)
  • (4)