accessibility menu, dialog, popup

UserName

Viewing profile as user:
GdBrBrBr

CAS RN 30010-20-9

IUPAC Name: gadolinium(3+) tribromide
Synonyms: gadolinium(III) bromide
Molecular Weight (g/mol): 396.96
Molecular Formula: Br3Gd
InChi Key: KGOKDPWKDBWITQ-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Gd+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results