accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3OOCH3P

CAS RN 30018-16-7

IUPAC Name: (1-ethoxy-1-oxopropan-2-yl)triphenylphosphanium bromide
Synonyms: (1-ethoxy-1-oxopropan-2-yl)triphenylphosphanium bromide
Molecular Weight (g/mol): 443.32
Molecular Formula: C23H24BrO2P
InChi Key: RSYXORMKBUFAMS-UHFFFAOYNA-M
SMILES: [Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results