accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNN

CAS RN 3002-78-6

IUPAC Name: 5-methyl-1,10-phenanthroline
Synonyms: 5-methyl-1,10-phenanthroline
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: UJAQYOZROIFQHO-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CN=C2C2=NC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results