accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHCl

CAS RN 300543-56-0

IUPAC Name: 1-[(4-chlorophenyl)(phenyl)methyl]piperazine
Synonyms: 1-[(4-chlorophenyl)(phenyl)methyl]piperazine
Molecular Weight (g/mol): 286.8
Molecular Formula: C17H19ClN2
InChi Key: UZKBSZSTDQSMDR-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(N1CCNCC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results