accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3006-96-0

IUPAC Name: 4-(hydroxymethyl)benzoic acid
Synonyms: p-hydroxymethyl benzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: WWYFPDXEIFBNKE-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)